Assay Design



Gene Transcripts

Gene Symbol
CRYGC
Entrez Gene ID
709553
Gene Name
crystallin, gamma C
Gene Aliases
EGK_04736
Chromosome Location
Chr.12: 95564939 - 95566628 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082964.1 NP_001076433.1 - 276 73
GenBank mRNA EF426305.1 - - 276 73

View Full Product Details