Assay Design
Gene Transcripts
- Gene Symbol
- CSH2
- Entrez Gene ID
-
717562
- Gene Name
- chorionic somatomammotropin hormone 2
- Gene Aliases
-
CSH-2, GHB2
- Chromosome Location
-
Chr.16: 60406860 - 60408561 on Build Mmul_10
- Gene Symbol
- LOC114669819
- Entrez Gene ID
-
114669819
- Gene Name
- somatotropin-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 60385391 - 60387217 on Build Mmul_10
- Gene Symbol
- LOC114673168
- Entrez Gene ID
-
114673168
- Gene Name
- somatotropin-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 60449639 - 60451321 on Build Mmul_10
- Gene Symbol
- LOC114673169
- Entrez Gene ID
-
114673169
- Gene Name
- somatotropin-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 60428239 - 60429940 on Build Mmul_10
View Full Product Details