Assay Design



Gene Transcripts

Gene Symbol
NPVF
Entrez Gene ID
613271
Gene Name
neuropeptide VF precursor
Gene Aliases
RFRP
Chromosome Location
Chr.3: 89901636 - 89905622 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033115.2 NP_001028287.2 - 587 106
GenBank mRNA EU834942.1 - - 587 106

View Full Product Details