Assay Design



Gene Transcripts

Gene Symbol
LCN9
Entrez Gene ID
711643
Gene Name
lipocalin 9
Gene Aliases
ESP17.4
Chromosome Location
Chr.15: 2596712 - 2600324 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171841.1 NP_001165312.1 - 300 106
XM_015116325.2 XP_014971811.2 - 358 106
GenBank mRNA AF218046.1 - - 300 106

View Full Product Details