Assay Design



Gene Transcripts

Gene Symbol
SCGB1C1
Entrez Gene ID
696344
Gene Name
secretoglobin family 1C member 1
Gene Aliases
EGK_05801
Chromosome Location
Chr.14: 182142 - 183534 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193437.1 NP_001180366.1 - 266 65

View Full Product Details