Assay Design



Gene Transcripts

Gene Symbol
OR4D9
Entrez Gene ID
699152
Gene Name
olfactory receptor 4D9
Gene Aliases
-
Chromosome Location
Chr.14: 14455893 - 14456843 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193500.1 NP_001180429.1 - 654 88

View Full Product Details