Assay Design



Gene Transcripts

Gene Symbol
LCN1
Entrez Gene ID
711974
Gene Name
lipocalin 1
Gene Aliases
-
Chromosome Location
Chr.15: 2724703 - 2729827 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194073.3 NP_001181002.3 - 250 83
XM_028834362.1 XP_028690195.1 - 552 83

View Full Product Details