Assay Design



Gene Transcripts

Gene Symbol
ZGLP1
Entrez Gene ID
712411
Gene Name
zinc finger GATA like protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 10058406 - 10063792 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194168.3 NP_001181097.2 - 1486 80
GenBank mRNA JV639511.1 - - 217 80

View Full Product Details