Assay Design



Gene Transcripts

Gene Symbol
COX4I2
Entrez Gene ID
713095
Gene Name
cytochrome c oxidase subunit IV isoform 2 (lung)
Gene Aliases
EGK_02535
Chromosome Location
Chr.10: 66874716 - 66884048 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194480.2 NP_001181409.1 - 49 76

View Full Product Details