Assay Design



Gene Transcripts

Gene Symbol
TRAPPC3L
Entrez Gene ID
715890
Gene Name
trafficking protein particle complex 3 like
Gene Aliases
BET3L
Chromosome Location
Chr.4: 20079093 - 20124149 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194514.3 NP_001181443.1 - 514 93

View Full Product Details