Assay Design



Gene Transcripts

Gene Symbol
COL10A1
Entrez Gene ID
716223
Gene Name
collagen type X alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.4: 19699478 - 19706652 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194546.1 NP_001181475.1 - 249 75

View Full Product Details