Assay Design



Gene Transcripts

Gene Symbol
GALNT4
Entrez Gene ID
100430612
Gene Name
polypeptide N-acetylgalactosaminyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.11: 89219361 - 89224752 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194761.2 NP_001181690.1 - 4642 92

View Full Product Details