Assay Design



Gene Transcripts

Gene Symbol
RNASEK
Entrez Gene ID
100423494
Gene Name
ribonuclease K
Gene Aliases
-
Chromosome Location
Chr.16: 6770268 - 6772403 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194804.1 NP_001181733.1 - 344 78
GenBank mRNA JV617285.1 - - 283 78
JV715476.1 - - 243 78

View Full Product Details