Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193354.1 | NP_001180283.1 | - | 153 | 95 |
XM_015131689.2 | XP_014987175.1 | - | 454 | 95 | |
XM_015131690.2 | XP_014987176.1 | - | 1960 | 95 | |
XM_015131691.2 | XP_014987177.1 | - | 283 | 95 | |
GenBank mRNA | JU320505.1 | - | - | 385 | 95 |