Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190904.1 | NP_001177833.1 | - | 1152 | 95 |
XM_015121339.2 | XP_014976825.1 | - | 1236 | 95 | |
GenBank mRNA | JU336796.1 | - | - | 1117 | 95 |
JU477555.1 | - | - | 1190 | 95 |