Assay Design



Gene Transcripts

Gene Symbol
CYP1D1
Entrez Gene ID
704920
Gene Name
cytochrome P450 1D1
Gene Aliases
-
Chromosome Location
Chr.15: 88448150 - 88466243 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246671.1 NP_001233600.1 - 985 92
GenBank mRNA GU289742.1 - - 985 92

View Full Product Details