Assay Design



Gene Transcripts

Gene Symbol
EIF4EBP2
Entrez Gene ID
100424761
Gene Name
eukaryotic translation initiation factor 4E binding protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 67363540 - 67387293 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257910.1 NP_001244839.1 - 458 80
GenBank mRNA JU333287.1 - - 458 80
JU474603.1 - - 497 80
JV046381.1 - - 438 80

View Full Product Details