Assay Design



Gene Transcripts

Gene Symbol
KCNIP3
Entrez Gene ID
704307
Gene Name
potassium voltage-gated channel interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.13: 303325 - 395630 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257864.1 NP_001244793.1 - 428 98
XM_015112830.2 XP_014968316.1 - 535 98
XM_015112831.2 XP_014968317.1 - 593 98
GenBank mRNA JU322491.1 - - 428 98
JU332335.1 - - 428 98
JU472856.1 - - 425 98
JU475363.1 - - 244 98
JU475364.1 - - 244 98
JV602086.1 - - 429 98

Gene Symbol
LOC114669762
Entrez Gene ID
114669762
Gene Name
calsenilin-like
Gene Aliases
-
Chromosome Location
Chr.13: 35817 - 44875 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028842728.1 XP_028698561.1 - 247 98

View Full Product Details