Assay Design



Gene Transcripts

Gene Symbol
PDP2
Entrez Gene ID
694851
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Gene Aliases
EGK_12867
Chromosome Location
Chr.20: 54057109 - 54063240 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260605.1 NP_001247534.1 - 1519 80
XM_015126322.2 XP_014981808.1 - 1712 80
XM_028840363.1 XP_028696196.1 - 1792 80
GenBank mRNA JU321275.1 - - 1514 80
JU472596.1 - - 1519 80
JV043271.1 - - 1629 80
JV043272.1 - - 1629 80
JV636774.1 - - 1633 80

View Full Product Details