Assay Design



Gene Transcripts

Gene Symbol
ENDOD1
Entrez Gene ID
699210
Gene Name
endonuclease domain containing 1
Gene Aliases
-
Chromosome Location
Chr.14: 88332897 - 88372835 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260700.1 NP_001247629.1 - 412 114
GenBank mRNA JU333531.1 - - 391 114
JV046506.1 - - 412 114

View Full Product Details