Assay Design



Gene Transcripts

Gene Symbol
EID1
Entrez Gene ID
714659
Gene Name
EP300 interacting inhibitor of differentiation 1
Gene Aliases
-
Chromosome Location
Chr.7: 25295100 - 25296820 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265838.2 NP_001252767.1 - 525 97
GenBank mRNA JU333754.1 - - 463 97
JV046627.1 - - 527 97
JV647155.1 - - 538 97

View Full Product Details