Assay Design



Gene Transcripts

Gene Symbol
PLSCR4
Entrez Gene ID
713430
Gene Name
phospholipid scramblase 4
Gene Aliases
-
Chromosome Location
Chr.2: 48354029 - 48420235 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266547.1 NP_001253476.1 - 269 69
GenBank mRNA JU335565.1 - - 301 69
JV045033.1 - - 269 69
JV656521.1 - - 299 69

View Full Product Details