Assay Design



Gene Transcripts

Gene Symbol
NDUFA2
Entrez Gene ID
697736
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Gene Aliases
-
Chromosome Location
Chr.6: 138026126 - 138028328 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261499.1 NP_001248428.1 - 167 130
GenBank mRNA JU322527.1 - - 302 130
JU473446.1 - - 292 130
JV045704.1 - - 167 130

View Full Product Details