Assay Design



Gene Transcripts

Gene Symbol
Clk2
Entrez Gene ID
365842
Gene Name
CDC-like kinase 2
Gene Aliases
-
Chromosome Location
Chr.2: 188476790 - 188488809 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014254.1 NP_001014276.1 6 - 7 942 86
XM_008761185.1 XP_008759407.1 6 - 7 1147 86
XM_008761186.1 XP_008759408.1 7 - 8 1014 86
XM_008761187.1 XP_008759409.1 6 - 7 1144 86
XM_008761188.1 XP_008759410.1 6 - 7 1126 86
XM_008761189.1 XP_008759411.1 6 - 7 1123 86
XM_008761190.1 XP_008759412.1 6 - 7 1123 86
XM_008761191.1 XP_008759413.1 6 - 7 1120 86
XM_008761192.1 XP_008759414.1 6 - 7 993 86
XM_008761193.1 XP_008759415.1 3 - 4 794 86
XM_008761194.1 XP_008759416.1 3 - 4 273 86
GenBank mRNA BC083788.1 - 6 - 7 942 86

View Full Product Details