Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030990.2 | NP_112252.1 | 3 - 4 | 603 | 67 |
XM_008773401.1 | XP_008771623.1 | 6 - 7 | 989 | 67 | |
GenBank mRNA | BC093596.1 | - | 2 - 3 | 464 | 67 |
FQ212680.1 | - | 2 - 3 | 317 | 67 | |
FQ213540.1 | - | 2 - 3 | 317 | 67 | |
FQ213548.1 | - | 2 - 3 | 317 | 67 | |
FQ213841.1 | - | 2 - 3 | 317 | 67 | |
FQ214012.1 | - | 2 - 3 | 317 | 67 | |
FQ214050.1 | - | 2 - 3 | 317 | 67 | |
FQ214153.1 | - | 2 - 3 | 317 | 67 | |
FQ214556.1 | - | 2 - 3 | 317 | 67 | |
M11185.1 | - | 3 - 4 | 578 | 67 | |
M25888.1 | - | 1 - 2 | 143 | 67 | |
X02809.1 | - | 3 - 4 | 507 | 67 |