Assay Design



Gene Transcripts

Gene Symbol
Trim8
Entrez Gene ID
688785
Gene Name
tripartite motif-containing 8
Gene Aliases
-
Chromosome Location
Chr.1: 266255797 - 266270694 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128083.1 NP_001121555.1 5 - 6 2062 100
GenBank mRNA CO393559.1 - 5 - 6 503 100

View Full Product Details