Assay Design



Gene Transcripts

Gene Symbol
Pbx3
Entrez Gene ID
311876
Gene Name
pre-B-cell leukemia homeobox 3
Gene Aliases
-
Chromosome Location
Chr.3: 13241031 - 13436084 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107834.1 NP_001101304.1 6 - 7 1052 88

View Full Product Details