Assay Design



Gene Transcripts

Gene Symbol
Tlx1
Entrez Gene ID
499361
Gene Name
T-cell leukemia, homeobox 1
Gene Aliases
RGD1563655
Chromosome Location
Chr.1: 264893162 - 264899424 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109166.1 NP_001102636.1 1 - 2 778 61

View Full Product Details