Assay Design



Gene Transcripts

Gene Symbol
Mapk8ip1
Entrez Gene ID
116457
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
Gene Aliases
JIP1, Jip-1, Mapk8ip
Chromosome Location
Chr.3: 81295023 - 81304181 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053777.1 NP_446229.1 8 - 9 1961 75
GenBank mRNA AF092450.1 - 7 - 8 1696 75
AF108959.1 - 7 - 8 1899 75
AF109772.1 - 7 - 8 1808 75
AF109773.1 - 7 - 8 1871 75
AF109774.1 - 8 - 9 1961 75
DQ377223.1 - 7 - 8 1771 75

View Full Product Details