Assay Design



Gene Transcripts

Gene Symbol
Acer2
Entrez Gene ID
313339
Gene Name
alkaline ceramidase 2
Gene Aliases
Asah3l
Chromosome Location
Chr.5: 105230538 - 105279732 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107943.1 NP_001101413.1 3 - 4 456 70
XM_006238363.2 XP_006238425.1 3 - 4 498 70
GenBank mRNA FQ229191.1 - 3 - 4 431 70

View Full Product Details