Assay Design



Gene Transcripts

Gene Symbol
Kcnh5
Entrez Gene ID
171146
Gene Name
potassium channel, voltage gated eag related subfamily H, member 5
Gene Aliases
Eag2
Chromosome Location
Chr.6: 97872831 - 98157087 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133610.2 NP_598294.1 7 - 8 1632 97
GenBank mRNA AF185637.1 - 7 - 8 1591 97
AJ250280.1 - 7 - 8 1634 97

View Full Product Details