Assay Design
Gene Transcripts
- Gene Symbol
- Slc6a15
- Entrez Gene ID
-
282712
- Gene Name
- solute carrier family 6 (neutral amino acid transporter), member 15
- Gene Aliases
-
Ntt73
- Chromosome Location
-
Chr.7: 45328207 - 45381920 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172321.1 |
NP_758824.1 |
8 - 9 |
2197 |
84 |
GenBank mRNA |
L22022.1 |
- |
8 - 9 |
2197 |
84 |
View Full Product Details