Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012619.2 | NP_036751.2 | 4 - 5 | 528 | 59 |
XM_008765215.1 | XP_008763437.1 | 2 - 3 | 468 | 59 | |
GenBank mRNA | BC078881.1 | - | 4 - 5 | 528 | 59 |
FQ219680.1 | - | 4 - 5 | 558 | 59 | |
M12337.1 | - | 4 - 5 | 513 | 59 |