Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012846.1 | NP_036978.1 | 3 - 4 | 542 | 79 |
XM_006254650.2 | XP_006254712.1 | 4 - 5 | 846 | 79 | |
XM_006254651.2 | XP_006254713.1 | 3 - 4 | 1663 | 79 | |
XM_006254652.2 | XP_006254714.1 | 3 - 4 | 863 | 79 | |
XM_006254653.2 | XP_006254715.1 | 3 - 4 | 698 | 79 | |
GenBank mRNA | X14232.1 | - | 3 - 4 | 542 | 79 |