Assay Design



Gene Transcripts

Gene Symbol
Cyp7a1
Entrez Gene ID
25428
Gene Name
cytochrome P450, family 7, subfamily a, polypeptide 1
Gene Aliases
CHAP, CYP7, CYP7S1
Chromosome Location
Chr.5: 19358734 - 19368431 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012942.2 NP_037074.1 1 - 2 115 81
GenBank mRNA J05430.1 - 1 - 2 110 81
J05460.1 - 1 - 2 116 81
J05509.1 - 1 - 2 127 81

View Full Product Details