Assay Design



Gene Transcripts

Gene Symbol
Hrh2
Entrez Gene ID
25461
Gene Name
histamine receptor H 2
Gene Aliases
H2R
Chromosome Location
Chr.17: 10912993 - 10931368 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012965.3 NP_037097.3 1 - 1 155 67
XM_008771486.1 XP_008769708.1 1 - 1 1462 67

View Full Product Details