Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013200.1 | NP_037332.1 | 7 - 8 | 816 | 75 |
XM_006242180.2 | XP_006242242.1 | 7 - 8 | 1272 | 75 | |
GenBank mRNA | BC085761.1 | - | 7 - 8 | 828 | 75 |
D43623.1 | - | 6 - 7 | 803 | 75 |