Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013221.2 | NP_037353.2 | 2 - 3 | 209 | 107 |
XM_006239975.2 | XP_006240037.1 | 2 - 3 | 272 | 107 | |
GenBank mRNA | BC091165.1 | - | 1 - 2 | 183 | 107 |
FQ223257.1 | - | 2 - 3 | 316 | 107 | |
U09551.1 | - | 1 - 2 | 204 | 107 |