Assay Design



Gene Transcripts

Gene Symbol
Gfap
Entrez Gene ID
24387
Gene Name
glial fibrillary acidic protein
Gene Aliases
-
Chromosome Location
Chr.10: 90990762 - 90999501 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017009.2 NP_058705.2 7 - 8 1161 76
GenBank mRNA BC088851.1 - 7 - 8 1161 76
L27219.1 - 7 - 8 1165 76
U03700.1 - 7 - 8 1185 76

View Full Product Details