Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017033.2 | NP_058729.2 | 9 - 10 | 1527 | 83 |
XM_006238442.2 | XP_006238504.1 | 10 - 11 | 1718 | 83 | |
XM_008763853.1 | XP_008762075.1 | 9 - 10 | 2070 | 83 | |
GenBank mRNA | BC099807.1 | - | 9 - 10 | 1527 | 83 |
BC128703.1 | - | 9 - 10 | 1532 | 83 | |
L11694.1 | - | 9 - 10 | 1510 | 83 |