Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017073.3 | NP_058769.4 | 1 - 2 | 105 | 63 |
GenBank mRNA | BC061559.1 | - | 1 - 2 | 118 | 63 |
BC072694.1 | - | 1 - 2 | 105 | 63 | |
FQ216093.1 | - | 1 - 2 | 110 | 63 | |
FQ227474.1 | - | 1 - 2 | 117 | 63 | |
FQ233770.1 | - | 1 - 2 | 117 | 63 | |
FQ234289.1 | - | 1 - 2 | 118 | 63 | |
M29579.1 | - | 1 - 2 | 116 | 63 | |
M91652.1 | - | 1 - 2 | 114 | 62 | |
X07921.1 | - | 1 - 2 | 116 | 63 |