Assay Design



Gene Transcripts

Gene Symbol
Bgn
Entrez Gene ID
25181
Gene Name
biglycan
Gene Aliases
BSPG1
Chromosome Location
Chr.X: 157319042 - 157331204 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017087.1 NP_058783.1 1 - 2 112 76
GenBank mRNA AY724513.1 - 1 - 2 115 76
BC072480.1 - 1 - 2 120 76
U17834.1 - 1 - 2 112 76

View Full Product Details