Assay Design
Gene Transcripts
- Gene Symbol
- Bmp3
- Entrez Gene ID
-
25667
- Gene Name
- bone morphogenetic protein 3
- Gene Aliases
-
PBMP3
- Chromosome Location
-
Chr.14: 12362912 - 12387572 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_017105.1 |
NP_058801.1 |
1 - 2 |
920 |
115 |
GenBank mRNA |
D63860.1 |
- |
1 - 2 |
920 |
115 |
View Full Product Details