Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017224.2 | NP_058920.1 | 6 - 7 | 1083 | 66 |
XM_006230978.2 | XP_006231040.1 | 6 - 7 | 1338 | 66 | |
GenBank mRNA | AB004559.1 | - | 6 - 7 | 1313 | 66 |
AF008221.1 | - | 6 - 7 | 1291 | 66 | |
BC104692.1 | - | 6 - 7 | 1083 | 66 |