Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017266.2 | NP_058962.2 | 4 - 5 | 523 | 92 |
XM_006229233.2 | XP_006229295.1 | 4 - 5 | 594 | 92 | |
XM_008759344.1 | XP_008757566.1 | 4 - 5 | 470 | 92 | |
GenBank mRNA | BC078862.1 | - | 4 - 5 | 523 | 92 |
JQ173140.1 | - | 4 - 5 | 511 | 92 | |
JQ409366.1 | - | 4 - 5 | 511 | 92 | |
U07177.1 | - | 3 - 4 | 424 | 92 |