Assay Design
Gene Transcripts
- Gene Symbol
- Ntm
- Entrez Gene ID
-
50864
- Gene Name
- neurotrimin
- Gene Aliases
-
Hnt, RNU16845
- Chromosome Location
-
Chr.8: 30039332 - 30479732 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_017354.1 |
NP_059050.1 |
6 - 7 |
1420 |
59 |
GenBank mRNA |
U16845.1 |
- |
6 - 7 |
1420 |
59 |
View Full Product Details