Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019125.1 | NP_061998.1 | - | 268 | 121 |
GenBank mRNA | BC062035.1 | - | - | 246 | 121 |
M27156.1 | - | - | 268 | 121 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008773239.1 | XP_008771461.1 | 2 - 3 | 368 | 121 |