Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019222.1 | NP_062095.1 | 9 - 10 | 1088 | 70 |
XM_006230773.2 | XP_006230835.1 | 9 - 10 | 1207 | 70 | |
XM_006230774.2 | XP_006230836.1 | 9 - 10 | 1172 | 70 | |
GenBank mRNA | AJ006064.1 | - | 9 - 10 | 1088 | 70 |
BC061558.1 | - | 8 - 9 | 1686 | 70 | |
FQ213235.1 | - | 5 - 6 | 461 | 70 |