Assay Design



Gene Transcripts

Gene Symbol
Pdp1
Entrez Gene ID
54705
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 1
Gene Aliases
Ppm2c
Chromosome Location
Chr.5: 25577593 - 25584327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019372.4 NP_062245.2 1 - 2 95 110
GenBank mRNA AF062740.1 - 1 - 2 77 110
FN800686.1 - 1 - 2 95 110

View Full Product Details