Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020092.1 | NP_064477.1 | 7 - 8 | 1087 | 82 |
XM_006231892.2 | XP_006231954.1 | 9 - 10 | 1606 | 82 | |
XM_006231894.2 | XP_006231956.1 | 8 - 9 | 6015 | 82 | |
GenBank mRNA | AF234765.1 | - | 7 - 8 | 1087 | 82 |